Learn More
Tris(2-thienyl)phosphine, 98+%, Thermo Scientific Chemicals
1840.00 SEK - 7045.00 SEK
Chemical Identifiers
CAS | 24171-89-9 |
---|---|
Molecular Formula | C12H9PS3 |
Molecular Weight (g/mol) | 280.36 |
MDL Number | MFCD00015471 |
InChI Key | KUCPTMZJPDVWJL-UHFFFAOYSA-N |
Synonym | tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine # |
PubChem CID | 90384 |
IUPAC Name | trithiophen-2-ylphosphane |
SMILES | S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1 |
Description
It employed as a intermediate for pharmaceutical and organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt employed as a intermediate for pharmaceutical and organic synthesis.
Solubility
Insoluble in water.
Notes
Air Sensitive, store away from air. Store in cool, dry place in tightly closed container. Store under dry inert gas.
Chemical Identifiers
24171-89-9 | |
280.36 | |
KUCPTMZJPDVWJL-UHFFFAOYSA-N | |
90384 | |
S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1 |
C12H9PS3 | |
MFCD00015471 | |
tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine # | |
trithiophen-2-ylphosphane |
Specifications
24171-89-9 | |
Recognizable | |
MFCD00015471 | |
Air Sensitive | |
Insoluble in water. | |
S1C=CC=C1P(C1=CC=CS1)C1=CC=CS1 | |
280.36 | |
280.37 | |
Tris(2-thienyl)phosphine |
28°C to 30°C | |
C12H9PS3 | |
203736 | |
tris 2-thienyl phosphine, phosphine, tri-2-thienyl, tri 2-thienyl phosphine, tri thiophen-2-yl phosphine, tri2-thienylphosphine, tris thiophen-2-yl phosphane, tri-2-thienylphosphine, trithienylphosphine, pubchem6427, tri 2-thienyl phosphine # | |
KUCPTMZJPDVWJL-UHFFFAOYSA-N | |
trithiophen-2-ylphosphane | |
90384 | |
≥98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 246-059-8
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only