missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSGA10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12593-1-AP-150UL
This item is not returnable.
View return policy
Description
TSGA10 may play a role in the sperm tail fibrous sheath, a major sperm tail structure.Specifications
TSGA10 | |
Polyclonal | |
Unconjugated | |
TSGA10 | |
CEP4L, testis specific, 10, TSGA10 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
211484, 252923, 80705 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6NY15, Q9BZW7, Q9Z220 | |
TSGA10 | |
TSGA10 Fusion Protein Ag3311 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |