missing translation for 'onlineSavingsMsg'
Learn More

TSGA10 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1656.73 SEK - 4721.67 SEK

Specifications

Antigen TSGA10
Concentration 0.13 mg/mL
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16850384
View Documents Promotion Details
Proteintech
12593-1-AP-20UL
20 μL
1656.73 SEK
20µL
Please to purchase this item. Need a web account? Register with us today!
16840384
View Documents Promotion Details
Proteintech
12593-1-AP-150UL
150 μL
4721.67 SEK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TSGA10 may play a role in the sperm tail fibrous sheath, a major sperm tail structure.
Specifications

Specifications

TSGA10
Western Blot, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Rat, Human, Mouse
Q6NY15, Q9BZW7, Q9Z220
211484, 252923, 80705
TSGA10 Fusion Protein Ag3311
Primary
-20°C
TSGA10
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CEP4L, testis specific, 10, TSGA10
TSGA10
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications