Learn More
LABPLAS Twirl'Em Sample Bags
Twirl'em sample bags are safe, flexible, contaminant-free and easy to use.
Brand: LABPLAS EPR-7012-FS
This item is not returnable.
View return policy
Description
- Twirl'Em sampling bags
- Safety tabs
- Clear, with writing area for easy labeling
- Closure with 1 round wire and 1 flat wire
- Available in various sizes
Specifications
Sample Bag | |
Clear with White Block | |
Sterile | |
Yes | |
178 x 305 mm | |
178 mm | |
305 mm |
Not Autoclavable | |
Single-use | |
76.2 μm | |
3 mil. | |
7 in. | |
12 in. |
Product Content Correction
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto