Learn More
LABPLAS Twirl'Em Sample Bags
Twirl'em sample bags are safe, flexible, contaminant-free and easy to use.
4027.17 SEK - 11745.00 SEK
Specifications
Product Type | Sample Bag |
---|---|
Autoclavable | Not Autoclavable |
Color | Clear with White Block |
Disposable | Single-use |
Sterility | Sterile |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | |||||
17950215
|
LABPLAS
EFR-1015S-FS |
254 x 381 mm |
11745.00 SEK
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17970295
|
LABPLAS
EPR-7012-FS |
178 x 305 mm | N/A | ||||||
Description
- Twirl'Em sampling bags
- Safety tabs
- Clear, with writing area for easy labeling
- Closure with 1 round wire and 1 flat wire
- Available in various sizes
Specifications
Sample Bag | |
Clear with White Block | |
Sterile |
Not Autoclavable | |
Single-use | |
Yes |