missing translation for 'onlineSavingsMsg'
Learn More

Emballator UN Approved Cans

Made of HDPE. Emballator UN Approved Cans approved by NET for storage and transport of dangerous goods.

Brand:  Emballator 48H0523-410-300

Encompass

 View more versions of this product

Product Code. 11304415

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Material: HDPE, approved for food contact
  • Color: Nature
  • UN Approved tamper evident closure
  • Approved by NET for storage and transport of dangerous goods
Specifications

Specifications

UN Approved Can
Natural
190 mm
306 mm
235 g
5 L
HDPE
130 mm
40 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.