Learn More
Fisherbrand™ Universal Stirrer Guide Replacement Lock Nut Pack
Description
- Consists of the following:
- Blue Lock Nut
- Ferrule
- Shaft Seal
Specifications
Fisherbrand™ Universal Stirrer Guide Replacement Lock Nut Pack | |
Blue Lock Nut, Ferrule & Shaft Seal | |
-200°C to +280°C |
12 mm Shaft | |
1 |