Learn More
Fisherbrand™ Universal Stirrer Guide Replacement Lock Nut Pack
Fisherbrand™ Universal Stirrer Guide Replacement Lock Nut Pack
298.00 SEK - 310.00 SEK
Specifications
Description | Fisherbrand™ Universal Stirrer Guide Replacement Lock Nut Pack |
---|---|
Includes | Blue Lock Nut, Ferrule & Shaft Seal |
Quantity | 1 |
Temperature (Metric) | -200°C to +280°C |
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
16266051
|
Fisherbrand™
5.06.RP |
6 mm Shaft |
298.00 SEK
Pack of 5 |
Estimated Shipment: 11-12-2024
Log in to see stock. |
|||||
16286051
|
Fisherbrand™
5.08.RP |
8 mm Shaft |
298.00 SEK
Pack of 5 |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
16226061
|
Fisherbrand™
5.12.RP |
12 mm Shaft |
310.00 SEK
Pack of 5 |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
16246061
|
Fisherbrand™
5.16.RP |
16 mm Shaft |
310.00 SEK
Pack of 5 |
Estimated Shipment: 20-12-2024
Log in to see stock. |
|||||
Description
- Consists of the following:
- Blue Lock Nut
- Ferrule
- Shaft Seal
Specifications
Fisherbrand™ Universal Stirrer Guide Replacement Lock Nut Pack | |
Blue Lock Nut, Ferrule & Shaft Seal | |
1 | |
-200°C to +280°C |