Learn More
Fisherbrand™ Universal Stirrer Guide Replacement Lock Nut Pack
Fisherbrand™ Universal Stirrer Guide Replacement Lock Nut Pack
Brand:  Fisherbrand™ 5.08.RP
 View more versions of this product
Product Code. 16286051
  • 298.00 SEK / Pack of 5
Estimated Shipment: 20-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Consists of the following:
  • Blue Lock Nut
  • Ferrule
  • Shaft Seal
Specifications
Specifications
Fisherbrand™ Universal Stirrer Guide Replacement Lock Nut Pack
Blue Lock Nut, Ferrule & Shaft Seal
-200°C to +280°C
8 mm Shaft
1