Learn More
uvex Unipur 6639 Polyurethane-Coated Assembly Gloves
Shop All Uvex Products
Click to view available options
Size:
6
8
9
10
11
Description
- Polyamide gloves with polyurethane coating on the palm and fingertips
- Coating offers a high level of abrasion resistance
- Enhanced grip capabilities in dry, or slightly damp applications
- Lightweight and sensitive, ideal for precision mechanical work
- Dark colored material hides dirt and stains
Specifications
Specifications
Coating Material | Polyurethane |
Color | Black |
Exterior Finish | Smooth |
Hand | Ambidextrous |
Material | Polyamide |
Product Type | Mechanical Protection Gloves |
Size | 9 |
Style | Full-hand Mitt |
EN388 Standard | 4131X |
Glove Cuff Style | Knit |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 11
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
uvex Unipur 6639 Polyurethane-Coated Assembly Gloves >
Spot an opportunity for improvement?Share a Content Correction