Learn More
Systec™ V-Series Vertical Floor-Standing Autoclaves, Systec VX
Sterilize solids, liquids and hazardous biological substances with these autoclaves. Systec™ V-Series Vertical Floor-Standing Autoclaves feature a compact and space-saving design with increased chamber height. For all laboratory applications including sophisticated sterilization processes.
Brand:  Systec™ 1095
 View more versions of this product
Product Code. 10009491
  • POA / Each
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Explore more special offers
Description
Description
Systec VX Vertical Top-loading Autoclave
Standard Features
  • Integrated, separate steam generator
  • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
  • Temperature and pressure range 140°C, 4 bar
  • LCD display and fully automatic microprocessor control
  • Number of sterilization programs: Up to 25
  • Code-secured access rights for changing parameters and further safety-relevant intervention
  • Internal memory for storing up to 500 sterilization cycles
  • Timer for starting programs
  • Autofill: automatic demineralized water feed for steam generation
  • Flexible PT-100 temperature sensor
  • Additional temperature sensor in condense exhaust
  • Temperature holding function for liquids after program finish
  • Special program for Durham tubes
  • Calculation of F0 value
  • Special program for waste sterilization with pulsed heat-up for more efficient air exhaust
  • Water-cooled steam exhaust, thermostatically controlled
  • Programmable automatic door-opening on completion of program
  • RS-232 and RS-485 interfaces for external data transmission (network-compatible)
Available options
  • Touch-Screen control
  • Extension of temperature and pressure ranges to 150°C / 5 bar (from chamber volume 65 liters and more)
Options for process optimization
  • Rapid cooling for efficient and safe cooling of liquids
  • Vacuum system for validatable sterilization of solids and waste materials in disposal bags
  • Superdry: for drying solids (only in combination with optional vacuum system)
  • Exhaust filtration (including condensate inactivation) for safe sterilization of hazardous biological substances
Options for documentation
  • Integrated printer for batch documentation
  • PC software for comprehensive documentation
  • Documentation SD: data storage on SD card for up to 10,000 sterilization cycles and transmission of data to a PC
  • Comlog: includes USB – and Ethernet-Connection and internal memory for up to 10,000 sterilization cycles, including documentation software on Comlog. The software can be called up independent of platform (PC, laptop, tablet, smart phone), facilitating remote service
  • AuditTrail: unalterable and traceable documentation acc. to FDA 21 CFR Part 11
Specifications
Specifications
780 mm
100 L (Total), 95 L (Nominal)
400 mm
1080 mm
Autoclave, Vertical
LCD display and fully automatic microprocessor control
4 bar
Top-loading
750 mm
For all laboratory applications even for sophisticated state-of-the-art sterilization processes
VX-95
140°C
380 to 400 V 50/60 Hz 16 A 3-phase plus neutral
550 mm