missing translation for 'onlineSavingsMsg'
Learn More

Systec™ V-Series Vertical Floor-Standing Autoclaves, Systec VX

Sterilize solids, liquids and hazardous biological substances with these autoclaves. Systec™ V-Series Vertical Floor-Standing Autoclaves feature a compact and space-saving design with increased chamber height. For all laboratory applications including sophisticated sterilization processes.

POA

Specifications

Design Top-loading
For Use With (Application) For all laboratory applications even for sophisticated state-of-the-art sterilization processes
Product Type Autoclave, Vertical
Temperature (Metric) 140°C
Display LCD display and fully automatic microprocessor control
View More Specs

Products 4
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
12022959
View accessory products View Documents
Systec™
1040
VX-40
POA
Each
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

10099481
View accessory products View Documents
Systec™
1065
VX-65
POA
Each
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

10367602
View accessory products View Documents
Systec™
1075
VX-75
POA
Each
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

10009491
View accessory products View Documents
Systec™
1095
VX-95
POA
Each
N/A
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

Systec VX Vertical Top-loading Autoclave

Standard Features
  • Integrated, separate steam generator
  • Internal heating elements within the autoclave chamber
  • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
  • Temperature and pressure range 140°C, 4 bar
  • LCD display and fully automatic microprocessor control
  • Number of sterilization programs: VX: up to 25; VE: 12; VB: 3
  • Code-secured access rights for changing parameters and further safety-relevant intervention
  • Internal memory for storing up to 500 sterilization cycles
  • Timer for starting programs
  • Autofill: automatic demineralized water feed for steam generation
  • Flexible PT-100 temperature sensor
  • Additional temperature sensor in condense exhaust
  • Temperature holding function for liquids after program finish
  • Special program for Durham tubes
  • Calculation of F0 value
  • Special program for waste sterilization with pulsed heat-up for more efficient air exhaust
  • Water-cooled steam exhaust, thermostatically controlled
  • Programmable automatic door-opening on completion of program
  • RS-232 and RS-485 interfaces for external data transmission (network-compatible)
 
Available options
  • Touch-Screen control
  • Extension of temperature and pressure ranges to 150°C / 5 bar (from chamber volume 65 liters and more)
Options for process optimization
  • Rapid cooling for efficient and safe cooling of liquids
  • Vacuum system for validatable sterilization of solids and waste materials in disposal bags
  • Superdry: for drying solids (only in combination with optional vacuum system)
  • Exhaust filtration (including condensate inactivation) for safe sterilization of hazardous biological substances
Options for documentation
  • Integrated printer for batch documentation
  • PC software for comprehensive documentation
  • Documentation SD: data storage on SD card for up to 10,000 sterilization cycles and transmission of data to a PC
  • Comlog: includes USB – and Ethernet-Connection and internal memory for up to 10,000 sterilization cycles, including documentation software on Comlog. The software can be called up independent of platform (PC, laptop, tablet, smart phone), facilitating remote service
  • AuditTrail: unalterable and traceable documentation acc. to FDA 21 CFR Part 11
Specifications

Specifications

Top-loading
Autoclave, Vertical
LCD display and fully automatic microprocessor control
For all laboratory applications even for sophisticated state-of-the-art sterilization processes
140°C
4 bar
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.