Learn More
Vaspin Mouse anti-Human, Unconjugated, Clone: 2A1B6, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67573-1-IG-20UL
 View more versions of this product
Product Code. 16800286
  • 1656.73 SEK / 20µL
Explore more special offers
This item is not returnable. View return policy
Description
Description
The Vaspin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Vaspin. This antibody recognizes human antigen. The Vaspin antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
Vaspin
Monoclonal
1000 μg/mL
Mouse
Primary
Human
Western Blot, ELISA
2A1B6
Unconjugated
20 μL
145264
IgG1