missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vaspin Mouse anti-Human, Unconjugated, Clone: 2A1B6, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | Vaspin |
---|---|
Clone | 2A1B6 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16800286
|
Proteintech
67573-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16890276
|
Proteintech
67573-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Vaspin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Vaspin. This antibody recognizes human antigen. The Vaspin antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Vaspin | |
1000 μg/mL | |
Monoclonal | |
Mouse | |
145264 | |
Primary |
2A1B6 | |
Western Blot, ELISA | |
Unconjugated | |
Human | |
IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title