Learn More
3M™ VFlex™ Particulate Respirator, FFP2, Unvalved
VFlex™ Particulate Respirators provide economic protection and have a compliable V-shaped pleat design with a 3M filter material that helps make breathing easier. Our respirators feature an embedded, adjustable metal nose clip for custom fit and side tabs for easier positioning on face.
Brand: 3M™ 7100199512
This item is not returnable.
View return policy
Description
- Economic respirator solution
- V-shaped pleats flex and expand around your mouth to aid comfort
- Proprietary 3M filter material helps you breathe easier
- Embossed front panel retains shape and keeps material away from the mouth
- Side tabs enable easier positioning
- Embedded, adjustable metal nose clip helps provide a custom and secure seal
- Flat-fold design makes it convenient and easy to store away
- Designed to work well with 3M Eyewear and Hearing Protection products
- Available in small and regular sizes to fit different face shapes
Specifications
White | |
Single-use | |
Particulate Respirator | |
No |
Flat-fold | |
FFP2 | |
Small |