Learn More
3M™ VFlex™ Particulate Respirator, FFP2, Unvalved
VFlex™ Particulate Respirators provide economic protection and have a compliable V-shaped pleat design with a 3M filter material that helps make breathing easier. Our respirators feature an embedded, adjustable metal nose clip for custom fit and side tabs for easier positioning on face.
Brand:  3M™ 7100199151
 View more versions of this product
Product Code. 17971030
  • 7755.00 SEK / Pack of 1000
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Economic respirator solution
  • V-shaped pleats flex and expand around your mouth to aid comfort
  • Proprietary 3M filter material helps you breathe easier
  • Embossed front panel retains shape and keeps material away from the mouth
  • Side tabs enable easier positioning
  • Embedded, adjustable metal nose clip helps provide a custom and secure seal
  • Flat-fold design makes it convenient and easy to store away
  • Designed to work well with 3M Eyewear and Hearing Protection products
  • Available in small and regular sizes to fit different face shapes
TRUSTED_SUSTAINABILITY
Specifications
Specifications
White
Single-use
Particulate Respirator
No
Flat-fold
FFP2
One Size