Learn More
MP Biomedicals Vitamin A Deficient Diet, Modified, Powder
Vitamin A deficient diet. Typically provides 3.89 kcal/gm.
Brand:  MP Biomedicals 0296021950
 View more versions of this product
Product Code. 11453011
  • 25008.58 SEK / 50kg
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • MP Bio's traditional vitamin A deficient diet, but revised to include essential trace minerals, DL-methionine and fiber
  • Typically provides 3.89 kcal/gm
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Autoclavable
Single-use
50 kg
Room Temperature
0.3048 m
Animal Diet
Powder
Non-sterile
1 ft.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.