Learn More
BRAND™ Watch Glass
Watch Glass constructed of soda-lime glass with ground edges.
Brand:  BRAND™ 150010
367.86 SEK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 10069980
  • 390.00 SEK / Pack of 10
In Stock
to see stock.
Explore more special offers
Description
  • Soda-lime glass
  • Ground edges
  • Stress-reduced to minimize risk of shattering
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Stress reduced edges
Soda Lime Glass
Ground
40 mm
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    22
    Hours
    13
    Minutes
    14
    Seconds
    53
Product Content Correction
Product Title
BRAND™ Watch Glass > Diameter: 40 mm