missing translation for 'onlineSavingsMsg'
Learn More

Karl Hecht™ Watch Glasses

These watch glasses are useful for evaporating liquids, holding solids, and heating small amounts of a sample.

Brand:  Karl Hecht™ 40010006

 View more versions of this product

Product Code. 11371395

  • 18.00 SEK / Each
Estimated Shipment: 26-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Acc. to DIN 12 341
  • With fused rim
  • Manufactured from glass
  • Available in different diameters
Specifications

Specifications

Fused rim
Round
6 cm
Glass
Evaporation, holding, heating
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Karl Hecht™ Watch Glasses > Dia: 6cm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.