Learn More
Bel-Art™ SP Scienceware™ Wilmad™ Spinner Bearing
Ensure shimming quality of your glass tubes. Bel-Art SP Scienceware™ Wilmad™ Spinner Bearing will determine which of your NMR tubes are warped and bent.
246.84 SEK - 259.00 SEK
Product Code | Brand | Model | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Diameter (Metric) Outer | Price | Quantity & Availability | ||||
15368608
|
Bel-Art™
66000-0042 |
SB-5-7 | For tubes 5mm O.D. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
15378608
|
Bel-Art™
66000-0043 |
SB-8-7 | For tubes 8mm O.D. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Useful in labs that wash, dry and reuse precision NMR tubes
- Warped tubes will bind in the spinner bearing while good tubes spin freely
- Keep a tube tester beside every NMR spectrometer for easy access
Specifications
Quartz, Supracil, or Precision tubes |