Learn More
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Inner | Price | Quantity & Availability | ||||||
13277169
|
Bohlender™
F718-04 |
1.6 mm |
222.00 SEK
Each |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
15318461
|
Bohlender™
F718-02 |
0.8 mm |
264.00 SEK
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
Description
- Connector made of PTFE
- For transition from Luer Lock connection to female thread UNF 1/4 in. 28G
- Inner cone of the Luer-Lock connection provides good sealing, the additional thread (either male or female) prevents accidental loosening
- Tubing is connected by means of tube end fittings to a female thread UNF 1/4 in. 28G
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bohlender™ Within Luer Lock PTFE Miniature Luer-Lock Connector