Learn More
Bohlender™ Within Luer Lock PTFE Miniature Luer-Lock Connector
Connect componets with Luer Lock such as syringes or cannulas to a tubing system.
222.00 SEK - 264.00 SEK
Products 2
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
13277169
View Documents
Bohlender™
F718-04
1.6 mm
222.00 SEK
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
15318461
View Documents
Bohlender™
F718-02
0.8 mm
264.00 SEK
Each
Estimated Shipment: 07-05-2025
to see stock.
Add to basket
Description
Description
  • Connector made of PTFE
  • For transition from Luer Lock connection to female thread UNF 1/4 in. 28G
  • Inner cone of the Luer-Lock connection provides good sealing, the additional thread (either male or female) prevents accidental loosening
  • Tubing is connected by means of tube end fittings to a female thread UNF 1/4 in. 28G
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bohlender™ Within Luer Lock PTFE Miniature Luer-Lock Connector