accessibility menu, dialog, popup

Test

Discover Our Special Offers

Save Now
2017-Fisher-Scientific-Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1082739-92-1

CAS RN 1082739-92-1

IUPAC Name: 3-(2H-1,3-benzodioxol-5-yl)-N-(1-phenylethyl)-[1,2]oxazolo[5,4-c]pyridin-5-amine
Synonyms: 3-(2H-1,3-benzodioxol-5-yl)-N-(1-phenylethyl)-[1,2]oxazolo[5,4-c]pyridin-5-amine
Molecular Weight (g/mol): 359.39
Molecular Formula: C21H17N3O3
InChi Key: DAAGOCSHXCDWDY-UHFFFAOYNA-N
SMILES: CC(NC1=NC=C2ON=C(C2=C1)C1=CC2=C(OCO2)C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products