man

Test

CAS RN 115-83-3

CH3OOOOCH3OOCH3OOCH3

CAS RN 115-83-3

IUPAC Name: 3-(octadecanoyloxy)-2,2-bis[(octadecanoyloxy)methyl]propyl octadecanoate
Synonyms: pentaerythrityl tetrastearate
Molecular Weight (g/mol): 1202.02
Molecular Formula: C77H148O8
InChi Key: OCKWAZCWKSMKNC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCC(=O)OCC(COC(=O)CCCCCCCCCCCCCCCCC)(COC(=O)CCCCCCCCCCCCCCCCC)COC(=O)CCCCCCCCCCCCCCCCC

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results