man

Test

CAS RN 1402-10-4

HOOOHHOHO

CAS RN 1402-10-4

IUPAC Name: (2S,3R,4R)-2-(hydroxymethyl)-3,4-dihydro-2H-pyran-3,4,5-triol
Synonyms: lichenin lichenan
Molecular Weight (g/mol): 162.14
Molecular Formula: C6H10O5
InChi Key: CXKKSSKKIOZUNR-ZLUOBGJFSA-N
SMILES: OC[C@@H]1OC=C(O)[C@H](O)[C@H]1O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results