CAS RN 156722-18-8
CAS RN 156722-18-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscience™ Rostafuroxin
Na+/K+ ATPase modulator; antihypertensive agent
2
3