Learn More
4-n-Hexylaniline, 98%, Thermo Scientific Chemicals
789.00 SEK - 2575.00 SEK
Chemical Identifiers
CAS | 33228-45-4 |
---|---|
Molecular Formula | C12H19N |
Molecular Weight (g/mol) | 177.291 |
MDL Number | MFCD00007927 |
InChI Key | OVEMTTZEBOCJDV-UHFFFAOYSA-N |
Synonym | p-hexylaniline, 4-n-hexylaniline, benzenamine, 4-hexyl, aniline, p-hexyl, 4-hexylbenzenamine, p-n-hexylaniline, unii-u3c0879zgc, 4-hexylphenylamine, 4-n-hexylaniline;, acmc-209hzh |
PubChem CID | 36382 |
IUPAC Name | 4-hexylaniline |
SMILES | CCCCCCC1=CC=C(C=C1)N |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11491140
|
Thermo Scientific Alfa Aesar
H26297.06 |
5 g |
789.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11401150
|
Thermo Scientific Alfa Aesar
H26297.14 |
25 g |
2575.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Hexylaniline is used in the fabrication of RP/ion-exchange, mixed-mode, monolithic materials for capillary LC. The product is dye stuff intermediate, yet it may use to manufacture the medicine
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids.
Chemical Identifiers
33228-45-4 | |
177.291 | |
OVEMTTZEBOCJDV-UHFFFAOYSA-N | |
36382 | |
CCCCCCC1=CC=C(C=C1)N |
C12H19N | |
MFCD00007927 | |
p-hexylaniline, 4-n-hexylaniline, benzenamine, 4-hexyl, aniline, p-hexyl, 4-hexylbenzenamine, p-n-hexylaniline, unii-u3c0879zgc, 4-hexylphenylamine, 4-n-hexylaniline;, acmc-209hzh | |
4-hexylaniline |
Specifications
33228-45-4 | |
283°C to 285°C | |
C12H19N | |
MFCD00007927 | |
p-hexylaniline, 4-n-hexylaniline, benzenamine, 4-hexyl, aniline, p-hexyl, 4-hexylbenzenamine, p-n-hexylaniline, unii-u3c0879zgc, 4-hexylphenylamine, 4-n-hexylaniline;, acmc-209hzh | |
OVEMTTZEBOCJDV-UHFFFAOYSA-N | |
4-hexylaniline | |
36382 | |
98% |
0.919 | |
>110°C (230°F) | |
1.525 | |
Light sensitive | |
Insoluble in water. | |
CCCCCCC1=CC=C(C=C1)N | |
177.291 | |
177.29 | |
4-n-Hexylaniline |
Safety and Handling
GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P330-P332+P313-P362-P501c
H302-H315-H319
missing translation for 'einecsNumber' : 251-409-8
missing translation for 'rtecsNumber' : BY2305000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.