Learn More
Avadomide, MedChemExpress
MedChemExpress Avadomide (CC 122) is an orally active cereblon modulator. Avadomide modulates cereblon E3 ligase activity and induces apoptosis of diffuse large B-cell lymphoma (DLBCL) cell lines. Avadomide exhibits potent antitumor and immunomodulatory activities.
Brand:  Medchem Express HY-100507
Encompass_Preferred
 View more versions of this product
Product Code. 16424356
  • 4014.63 SEK / 100mg
Explore more special offers
Description
Description
Avadomide (CC 122) is an orally active cereblon modulator. Avadomide modulates cereblon E3 ligase activity and induces apoptosis of diffuse large B-cell lymphoma (DLBCL) cell lines. Avadomide exhibits potent antitumor and immunomodulatory activities.
Specifications
Specifications
Avadomide
Gray
C14H14N4O3
CC 122
O=C(C(N1C(C)=NC2=C(C(N)=CC=C2)C1=O)CC3)NC3=O
286.29
Research
Cancer-programmed cell death
1015474-32-4
Research
100 mg
DMSO : 33.33 mg/mL (116.42 mM; Need ultrasonic)
286.29
99.5%
Solid
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Avadomide, MedChemExpress