Learn More
MK-5108, MedChemExpress
MedChemExpress MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.
Brand: Medchem Express HY-13252
Description
MK-5108 is a highly potent and specific inhibitor of Aurora A kinase with an IC50 value of 0.064 nM.Specifications
MK-5108 | |
White | |
C22H21ClFN3O3S | |
VX-689 | |
FC(C(Cl)=CC=C1)=C1O[C@H](CC2)CC[C@@]2(C(O)=O)CC3=CC=CC(NC4=NC=CS4)=N3 | |
461.94 | |
Research | |
Cancer-Kinase/protease |
1010085-13-8 | |
Research | |
5 mg | |
DMSO : 12.5 mg/mL (27.06 mM; Need ultrasonic) | |
461.94 | |
96.69% | |
Powder |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only