Learn More
Carbendazim, MedChemExpress
MedChemExpress Carbendazim is a potent and orally active broad-spectrum benzimidazole fungicide and can be acts as a pesticide for fungal diseases research, such as Seproria, Fusarium and Sclerotina. Carbendazim is a benzimidazole (HY-Y1825) derivative with antitumor activity and used for cancer research, especially advanced solid tumors and lymphoma.
Brand:  Medchem Express HY-13582
Encompass_Preferred
 View more versions of this product
Product Code. 16451676
  • 1198.17 SEK / 500mg
Explore more special offers
Description
Description
Carbendazim is a potent and orally active broad-spectrum benzimidazole fungicide and can be acts as a pesticide for fungal diseases research, such as Seproria, Fusarium and Sclerotina. Carbendazim is a benzimidazole (HY-Y1825) derivative with antitumor activity and used for cancer research, especially advanced solid tumors and lymphoma.
Specifications
Specifications
Carbendazim
10605-21-7
500 mg
C9H9N3O2
DMSO : 6.8 mg/mL (35.57 mM; Need ultrasonic)
191.19
99.81%
Solid
White
Research
COVID-19-immunoregulation
O=C(OC)NC1=NC2=CC=CC=C2N1
191.19
Research
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Carbendazim, MedChemExpress