Learn More
Invitrogen™ Cu/ZnSOD Human Matched Antibody Pair
Matched Antibody Pair
Brand:  Invitrogen™ BMS222MST
Product Code. 15551737
  • 7340.00 SEK / Pack of 10
Explore more special offers
Includes:
  • 1 vial (2.75mL) monoclonal Coating Antibody to human Cu/ZnSOD (100μg/mL)
  • 1 vial (55μL) HRP-Conjugate anti-human Cu/ZnSOD monoclonal antibody
  • 1 vial (60μL) human Cu/ZnSOD Standard protein, 400ng/mL
This item is not returnable. View return policy
Description
Description
The Human Cu/ZnSOD Matched Antibody Pair Kit comes with pre-matched antibody pairs and standards to develop your own enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of human Cu/ZnSOD. Buffer reagents and microplates needed to complete the ELISA reaction are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate,which is an overnight process. Samples,standards,or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody,a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
SOD1 (superoxide dismutase) binds copper and zinc ions and is one of two isozymes responsible for destroying free superoxide radicals in the body. This isozyme is a soluble cytoplasmic protein, acting as a homodimer to convert naturally-occuring but harmful superoxide radicals to molecular oxygen and hydrogen peroxide. The other isozyme is a mitochondrial protein. Mutations in this gene have been implicated as causes of familial amyotrophic lateral sclerosis. Rare transcript variants have been reported for this gene.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cu/ZnSOD
Monoclonal
P00441
SOD1
RUO
6647
-20°C
ELISA
HRP
Cu/Zn Superoxide Dismutase
10 x 96 Tests
Primary
Human
ELISA Kit
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Cu/ZnSOD Human Matched Antibody Pair > 10 x 96 Tests