missing translation for 'onlineSavingsMsg'
Learn More

Eurofins DiscoverX DMPK Protein, Active

Active, 6His-tagged, recombinant, human DMPK, amino acids 1-549, for use in Kinase Assays.

Encompass_Preferred

6046.30 SEK - 46147.75 SEK

Specifications

Formulation mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Molecular Weight (g/mol) 65 kDa
Species Human
Recombinant Recombinant
Protein Tag N-terminal 6His
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17340494
Encompass_Preferred
View Documents
Eurofins DiscoverX
14-649
10 μg
6046.30 SEK
10µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17350494
Encompass_Preferred
View Documents
Eurofins DiscoverX
14-649M
250 μg
46147.75 SEK
250µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Myotonic dystrophy (DM1) is the most common form of muscular dystrophy in adults, caused by amplification of an unstable (CTG)n repeat in the 3' untranslated region (3'-UTR) of the DM1 protein kinase (DMPK) gene. Myotonic dystrophy protein kinase (DMPK) is a serine threonine kinase encoded by the DM-1 locus.

GenBank NM_004409

Specifications

Specifications

mg/mL of enzyme in 50 mM Tris/HCl pH 7.5, 300 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Human
N-terminal 6His
Truncated
DMPK, active
65 kDa
Recombinant
Recombinant enzyme expressed in Sf21 insect cells
≥66%
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Eurofins DiscoverX DMPK Protein, Active

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.