missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Fluvoxamine maleate Product Code.: 15892474
compliance-icons

Thermo Scientific Chemicals Fluvoxamine maleate

Product Code. 15892474
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 15892474

Brand: Thermo Scientific Chemicals 462350010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61718-82-9
Molecular Formula C19H25F3N2O6
Molecular Weight (g/mol) 434.41
MDL Number MFCD00269809,MFCD00269809
InChI Key LFMYNZPAVPMEGP-PIDGMYBPSA-N
IUPAC Name (2Z)-but-2-enedioic acid; (E)-(2-aminoethoxy)({5-methoxy-1-[4-(trifluoromethyl)phenyl]pentylidene})amine
SMILES OC(=O)\C=C/C(O)=O.COCCCC\C(=N/OCCN)C1=CC=C(C=C1)C(F)(F)F

Specifications

Melting Point 120.0°C to 122.0°C
Color Clear
Quantity 1 g
Solubility Information Solubility aq. soln.: slightly soluble. Other solubilities: very soluble in chloroform and ethanol, soluble in methanol, insoluble in ether
Formula Weight 434.41
Chemical Name or Material Fluvoxamine maleate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466822 Certificate TypeCertificate of Analysis Date31/10/2024 Catalog Number462350010,462350050
Lot NumberA0386105 Certificate TypeCertificate of Analysis Date20/09/2022 Catalog Number462350010,462350050
Lot NumberA0422014 Certificate TypeCertificate of Analysis Date27/08/2020 Catalog Number462350010,462350050
compliance-icons
Product Identifier
  • Fluvoxamine maleate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Fluvoxamine maleate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Fluvoxamine maleate >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.