Learn More
Glass Syringes
Use these syringes for a variety of applications. Glass syringes feature a metal tip and are fully interchangeable.
Brand: S Murray SAMCO G812/04
Description
- Glass syringe with metal tip
- Luer fitting
- Fully interchangeable
- Available in a variety of sizes
Specifications
Glass | |
Syringe | |
10 mL |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
Glass Syringes > Volume: 10mL
Spot an opportunity for improvement?Share a Content Correction