Learn More
Granisetron Hydrochloride, MedChemExpress
MedChemExpress Granisetron (Hydrochloride) (BRL 43694A) is a serotonin 5-HT3 receptor antagonist used as an antiemetic to treat nausea and vomiting following chemotherapy.
Brand: Medchem Express HY-B0071A

Description
Granisetron (Hydrochloride) (BRL 43694A) is a serotonin 5-HT3 receptor antagonist used as an antiemetic to treat nausea and vomiting following chemotherapy.Specifications
Granisetron Hydrochloride | |
White | |
C18H25ClN4O | |
BRL 43694A | |
CN1N=C(C(N[C@H]2C[C@H](N3C)CCC[C@H]3C2)=O)C4=CC=CC=C41.[H]Cl | |
348.87 | |
Research | |
Neuroscience-Neuromodulation |
107007-99-8 | |
Research | |
100 mg | |
H2O : 100 mg/mL (286.64 mM; Need ultrasonic) ∣DMSO : 7.69 mg/mL (22.04 mM; Need ultrasonic) | |
348.87 | |
99.9% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only