missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies GSA 10

Hedgehog pathway activator; Activates Smoothened (SMO)

POA - 13464.90 SEK

Specifications

CAS 300833-95-8
Molecular Formula C26H30N2O5
Purity ≥95%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17140182
View Documents Promotion Details
STEMCELL Technologies
73172
10 mg
POA
10mg
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

17150182
View Documents Promotion Details
STEMCELL Technologies
73174
50 mg
13464.90 SEK
50mg
View alternative products
Save Up to 
»
Estimated Shipment: 16-08-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • GSA 10 is an agonist of Smoothened (SMO), a cell surface receptor and mediator of the hedgehog signaling pathway
  • It is a quinolinecarboxamide derivative that binds in a distinct binding pocket from cyclopamine
  • GSA10 is active at an EC50 of 1.2 μM in an alkaline phosphatase-based mouse mesenchymal cell line (C3H10T1/2) differentiation assay (Gorojankina et al.)
  • Promotes differentiation of multipotent mesenchymal progenitor cells into osteoblasts (Gorojankina et al.)
Specifications

Specifications

300833-95-8
C26H30N2O5
≥95%
Videos
SDS
Documents

Documents

Product Certifications