Learn More
Lazabemide, MedChemExpress
MedChemExpress Lazabemide (Ro 19-6327) is a selective, reversible inhibitor of monoamine oxidase B (MAO-B) (IC50=0.03 μM) but less active for MAO-A (IC50>100 μM). Lazabemide inhibits monoamine uptake at high concentrations, the IC50 values are 86 μM, 123 μM and >500 μM for noradrenalin, serotonin and dopamine uptake, respectively. Lazabemide can be used for the research of parkinson and alzheimer′s disease.
Brand: Medchem Express HY-14201
Description
Lazabemide (Ro 19-6327) is a selective, reversible inhibitor of monoamine oxidase B (MAO-B) (IC50=0.03 μM) but less active for MAO-A (IC50>100 μM). Lazabemide inhibits monoamine uptake at high concentrations, the IC50 values are 86 μM, 123 μM and >500 μM for noradrenalin, serotonin and dopamine uptake, respectively. Lazabemide can be used for the research of parkinson and alzheimer′s disease.Specifications
Lazabemide | |
Light Yellow | |
C8H10ClN3O | |
Ro 19-6327 | |
ClC1=CC=C(C(NCCN)=O)N=C1 | |
199.64 | |
Research | |
Neuroscience-Neuromodulation |
103878-84-8 | |
Research | |
10 mg | |
DMSO : 5 mg/mL (25.05 mM; Need ultrasonic) | |
199.64 | |
99.76% | |
Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only