Learn More
Tirasemtiv, MedChemExpress
MedChemExpress Tirasemtiv is an activator of the fast skeletal muscle troponin complex.
Brand: Medchem Express HY-15964

Description
Tirasemtiv is an activator of the fast skeletal muscle troponin complex.Specifications
Tirasemtiv | |
1005491-05-3 | |
100 mg | |
C12H14N4O | |
CK-2017357 | |
O=C1N(C(CC)CC)C2=NC(C#C)=CN=C2N1 | |
230.27 | |
Research |
Solid | |
Gray | |
Research | |
Neuroscience-Neuromodulation | |
DMSO : 10.91 mg/mL (47.38 mM; Need ultrasonic) | |
230.27 | |
99.83% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only