Learn More
Lifitegrast, MedChemExpress
MedChemExpress Lifitegrast (SAR 1118) is a potent integrin antagonist. Lifitegrast blocks the binding of intercellular adhesion molecule 1 (ICAM-1) to lymphocyte function-associated antigen 1 (LFA-1), interrupting the T cell-mediated inflammatory cycle. Lifitegrast inhibits Jurkat T cell attachment to ICAM-1 with an IC50 of 2.98 nM. Lifitegrast can be used for researching dry eye disease.
Brand: Medchem Express HY-19344
Description
Lifitegrast (SAR 1118) is a potent integrin antagonist. Lifitegrast blocks the binding of intercellular adhesion molecule 1 (ICAM-1) to lymphocyte function-associated antigen 1 (LFA-1), interrupting the T cell-mediated inflammatory cycle. Lifitegrast inhibits Jurkat T cell attachment to ICAM-1 with an IC50 of 2.98 nM. Lifitegrast can be used for researching dry eye disease.Specifications
Lifitegrast | |
Off-White | |
C29H24Cl2N2O7S | |
SAR 1118 SHP-606 | |
O=C(O)[C@H](CC1=CC=CC(S(=O)(C)=O)=C1)NC(C2=C(Cl)C3=C(CN(C(C4=CC=C5C=COC5=C4)=O)CC3)C=C2Cl)=O | |
615.48 | |
Research | |
COVID-19-immunoregulation |
1025967-78-5 | |
Research | |
50 mg | |
DMSO : ≥ 29 mg/mL (47.12 mM) | |
615.48 | |
99.09% | |
Solid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only