missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRT 68921 dihydrochloride
ULK inhibitor
1620.00 SEK - 5340.00 SEK
Specifications
Chemical Name or Material | N-[3-[[5-Cyclopropyl-2-[(1,2,3,4-tetrahydro-2-methyl-6-isoquinolinyl)amino]-4-pyrimidinyl]amino]propyl]cyclobutanecarboxamide dihydrochloride |
---|---|
Assay Percent Range | >98% |
Molecular Formula | C25H34N6O.2HCl |
Formula Weight | Observed MW: 507.5 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18728649
|
Tocris Bioscience
5780/25 |
25 mg |
5340.00 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18799817
|
Tocris Bioscience
5780/5 |
5 mg |
1620.00 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18789817
|
Tocris Bioscience
5780/5 |
5 mg |
1620.00 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRT 68921 dihydrochloride is a potent ULK inhibitor (IC50 values are 1.1 and 2.9 nM for ULK2 and ULK1, respectively). Inhibits autophagy in mouse embryonic fibroblasts.Specifications
N-[3-[[5-Cyclopropyl-2-[(1,2,3,4-tetrahydro-2-methyl-6-isoquinolinyl)amino]-4-pyrimidinyl]amino]propyl]cyclobutanecarboxamide dihydrochloride | |
>98% | |
C25H34N6O.2HCl | |
Observed MW: 507.5 |
Safety and Handling
Recommended Storage : Desiccate at Room Temperature
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
MRT 68921 dihydrochloride