Learn More
Thermo Scientific™ Nunc™ Cell Factory™ System
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems.
2213.36 SEK - 19505.00 SEK
Specifications
Description | Original Cell Factory |
---|---|
Product Type | Cell Factory System |
Green Features | Less waste, renewable material |
Product Code | Brand | Tray Layers | Culture Area | No. per Case | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tray Layers | Culture Area | No. per Case | Price | Quantity & Availability | ||||||
10344321
![]() |
Thermo Scientific™
139446 |
40 | 25,280 cm² | 2/Cs. |
19505.00 SEK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
10113922
![]() |
Thermo Scientific™
140004 |
4 | 2528 cm² | 10/Cs. |
14630.00 SEK
Pack of 10 |
In Stock
Log in to see stock. |
||||||
12677093
|
Thermo Scientific™
140004 |
4 | 2528 cm² | 10/Cs. | N/A | N/A | N/A | |||||
10049040
|
Thermo Scientific™
164327 |
10 | 6320 cm² | 2/Cs. |
5230.00 SEK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
10634551
![]() |
Thermo Scientific™
165250 |
1 | 632 cm² | 8/Cs. |
5610.00 SEK
Pack of 8 |
In Stock
Log in to see stock. |
||||||
10394221
|
Thermo Scientific™
167695 |
2 | 1264 cm² | 6/Cs. |
4320.00 SEK
Pack of 6 |
In Stock
Log in to see stock. |
||||||
13101594
|
Thermo Scientific™
167695 |
2 | 1264 cm² | 6/Cs. | N/A | N/A | N/A | |||||
10604901
![]() |
Thermo Scientific™
170009 |
10 | 6320 cm² | 6/Cs. |
14310.00 SEK
Pack of 6 |
In Stock
Log in to see stock. |
||||||
Description
Obtain faster results and lot-to-lot consistency with Thermo Scientific™ Nunc™ Cell Factory™ systems. A proven solution for large-scale production of cells, vaccines and therapeutic proteins, these systems have the same growth kinetics as laboratory-scale cell culture product. The ports of the system make it easy to customize and close, with custom tubing assemblies that facilitate venting, filling and harvesting. Available in 1-, 2-, 4-, 10- and 40-tray versions.
- Ideal for adherent cells
- Boost production efficiency with more surface area in a small footprint
- Reduce contamination risks with a closed system
Specifications
Original Cell Factory | |
Cell Factory System | |
Less waste, renewable material |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.