Learn More
Thermo Scientific™ Automation Tips for Beckman™ Liquid Handling Systems with AP96 Dispense Head
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems through the superior tip straightness and low %CV.
1585.00 SEK - 12460.00 SEK
Specifications
Color | Clear |
---|---|
Certifications/Compliance | ISO 9001 and ISO 13485 |
Format | Rack |
Product Code | Brand | Volume (Metric) | Filtered | For Use With (Equipment) | Fit for Purpose | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Filtered | For Use With (Equipment) | Fit for Purpose | Price | Quantity & Availability | ||
15292929
|
Thermo Scientific™
912-021 |
8 μL | Filtered | Beckman Coulter Liquid Handling Systems | Sterile & PCR Ready |
9980.00 SEK
Pack of 4800 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||
15202949
|
Thermo Scientific™
912-261 |
10 μL | Non-Filtered | Beckman Coulter Liquid Handling Systems | Sterile |
7115.00 SEK
Pack of 4800 |
Estimated Shipment: 24-01-2025
Log in to see stock. |
||
15212949
|
Thermo Scientific™
912-262 |
10 μL | Non-Filtered | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96 | Certified Quality |
6440.00 SEK
Pack of 4800 |
Estimated Shipment: 17-01-2025
Log in to see stock. |
||
16456939
|
Thermo Scientific™
916-021 |
40 μL | Filtered | Beckman Coulter Liquid Handling Systems | Sterile & PCR Ready |
9980.00 SEK
Pack of 4800 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||
16466939
|
Thermo Scientific™
916-261 |
50 μL | Non-Filtered | Beckman Coulter Liquid Handling Systems | Sterile |
5850.00 SEK
Pack of 4800 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||
15105393
|
Thermo Scientific™
916-262 |
50 μL | Non-Filtered | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96 | Certified Quality |
5220.00 SEK
Pack of 4800 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||
12578468
|
Thermo Scientific™
916-262-05 |
50 μL | Non-Filtered | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96 | Certified Quality |
6155.00 SEK
Pack of 4800 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||
11861723
|
Thermo Scientific™
918-021 |
20 μL | Filtered | Beckman Coulter Liquid Handling Systems | Sterile & PCR Ready |
9980.00 SEK
Pack of 4800 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||
11761273
|
Thermo Scientific™
918-021-05 |
20 μL | Filtered | Beckman Coulter Liquid Handling Systems | Sterile & PCR Ready |
12460.00 SEK
Pack of 4800 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||
11811733
|
Thermo Scientific™
918-261 |
20 μL | Non-Filtered | Beckman Coulter Liquid Handling Systems | Sterile |
5850.00 SEK
Pack of 4800 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||
11741273
|
Thermo Scientific™
918-261-05 |
20 μL | Non-Filtered | Beckman Coulter Liquid Handling Systems | Sterile |
7080.00 SEK
Pack of 4800 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||
11851723
|
Thermo Scientific™
918-262 |
20 μL | Non-Filtered | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96 | Certified Quality |
5220.00 SEK
Pack of 4800 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||
11751273
|
Thermo Scientific™
918-262-05 |
20 μL | Non-Filtered | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96 | Certified Quality |
6275.00 SEK
Pack of 4800 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||
11801733
|
Thermo Scientific™
919-021 |
130 μL | Filtered | Beckman Coulter Liquid Handling Systems | Sterile & PCR Ready |
9980.00 SEK
Pack of 4800 |
Estimated Shipment: 31-07-2024
Log in to see stock. |
||
11791273
|
Thermo Scientific™
919-021-05 |
130 μL | Filtered | Beckman Coulter Liquid Handling Systems | Sterile & PCR Ready |
12250.00 SEK
Pack of 4800 |
In Stock
Log in to see stock. |
||
16476939
|
Thermo Scientific™
919-021G |
130 μL | Filtered | Beckman Coulter Liquid Handling Systems | Sterile & PCR Ready |
9980.00 SEK
Pack of 4800 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||
11881723
|
Thermo Scientific™
919-261 |
250 μL | Non-Filtered | Beckman Coulter Liquid Handling Systems | Sterile |
5950.00 SEK
Pack of 4800 |
In Stock
Log in to see stock. |
||
11771273
|
Thermo Scientific™
919-261-05 |
250 μL | Non-Filtered | Beckman Coulter Liquid Handling Systems | Sterile |
7080.00 SEK
Pack of 4800 |
Estimated Shipment: 26-11-2024
Log in to see stock. |
||
16486939
|
Thermo Scientific™
919-261G |
250 μL | Non-Filtered | Beckman Coulter Liquid Handling Systems | Sterile |
5850.00 SEK
Pack of 4800 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||
11891723
|
Thermo Scientific™
919-262 |
250 μL | Non-Filtered | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96 | Certified Quality |
5220.00 SEK
Pack of 4800 |
In Stock
Log in to see stock. |
||
11781273
|
Thermo Scientific™
919-262-05 |
250 μL | Non-Filtered | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96 | Certified Quality |
6275.00 SEK
Pack of 4800 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||
16496939
|
Thermo Scientific™
919-262G |
250 μL | Non-Filtered | Beckman Coulter Biomek FX, Beckman Coulter Biomek NX, Beckman Coulter Biomek 3000, Beckman Coulter Multimek AP96 | Certified Quality |
5220.00 SEK
Pack of 4800 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||
11741353
|
Thermo Scientific™
2760 |
50 μL | Filtered | Universal Fit Pipettor | Sterile |
7790.00 SEK
Pack of 4800 |
In Stock
Log in to see stock. |
||
11515514
|
Thermo Scientific™
2790 |
175 μL | Filtered | Beckman Coulter Liquid Handling Systems | Sterile |
1585.00 SEK
Pack of 960 |
Estimated Shipment: 20-12-2024
Log in to see stock. |
||
15272289
|
Thermo Scientific™
3762 |
50 μL | Non-Filtered | Beckman Coulter Multimek Type II Head | Certified Quality |
4050.00 SEK
Pack of 4800 |
Estimated Shipment: 12-11-2024
Log in to see stock. |
||
Description
Increase pipetting performance with Thermo Scientific™ Sterile Pipette Tips for Beckman™ Liquid Handling Systems, which undergo a rigorous 15-point quality control program to ensure consistency. These automation tips provide accurate and precise performance due to superior tip straightness and a low coefficient of variation. The pipette tips are validated on the actual workstations for which they are created.
Features:- Available with Thermo Scientific™ ART™ filter technology, low retention and extended length
- DNase, RNase, DNA, ATP, PCR Inhibitor, and Endotoxin Free
Specifications
Clear | |
ISO 9001 and ISO 13485 | |
Rack |