Learn More
PDE-9 inhibitor, MedChemExpress
MedChemExpress PDE-9 inhibitor is useful for neurodegenerative diseases.
Brand: Medchem Express HY-50865
This item is not returnable.
View return policy
Description
PDE-9 inhibitor is useful for neurodegenerative diseases.Specifications
PDE-9 inhibitor | |
1082743-70-1 | |
50 mg | |
C22H27N5O2 | |
DMSO : ≥ 100 mg/mL (254.14 mM) | |
393.48 | |
99.83% |
Solid | |
White | |
Research | |
Neuroscience-Neurodegeneration | |
O=C1NC([C@H]2[C@@H](CN(C2)CC3=CC=CC=C3)C)=NC4=C1C=NN4C5CCOCC5 | |
393.48 | |
Research |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
PDE-9 inhibitor, MedChemExpress