Description
- Black polyamide knitted glove
- Black oil proof nitrile coating on palm and fingertips
- The knitting technique helps to obtain a glove that conforms snugly to the hand, with good dexterity
- The glove offers maximum comfort and added sturdiness
- The tight knit gives the glove perfect support, limits the penetration of dirt and offers a good touch
- The ventilated back and PU coating optimise the glove's breathability
- The knitted and elasticated wrist gives the glove good support
- The filament yarn reduce linting for the most sensitive applications and easily laundered for extended use
- The nitrile coating at the palm and fingertips provides outstanding abrasion resistance and impermeability to oils
- Its dark color makes it more soilproof, thus ensuring a longer life
Specifications
Specifications
Certifications/Compliance | EN 420-03 Basic standards, EN 388-16 Mechanical hazards |
Description | 13 gauge |
Coating Material | Polyamide, Nitrile |
Color | Black |
Size | 9 |
Length (Metric) | 23 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Honeywell™ POLYTRIL™ Black Gloves >
Spot an opportunity for improvement?Share a Content Correction