Learn More
Eurofins DiscoverX Pyk2 Protein, Active
Active, N-Terminal His6-tagged, recombinant, full-length, human Pyk2, for use in Kinase Assays.

6046.30 SEK - 46147.75 SEK
Specifications
Formulation | mg/mL of enzyme in Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. |
---|---|
Molecular Weight (g/mol) | 117.2 kDa |
Species | Human |
Recombinant | Recombinant |
Protein Tag | N-terminal 6His |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17360354
![]() |
Eurofins DiscoverX
14-567 |
10 μg |
6046.30 SEK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17370354
![]() |
Eurofins DiscoverX
14-567M |
250 μg |
46147.75 SEK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Proline-rich tyrosine kinase 2 (PTK2B) (also called CAKb for cell adhesion kinase b, RAFTK for related adhesion focal tyrosine kinase, or CADTK for calcium-dependent tyrosine kinase) is a non-receptor tyrosine kinase structurally related to focal adhesion kinase (FAK). PTK2B interacts with Src family kinases and focal adhesion proteins such as p130Crk-associated substrate (p130Cas) and its homologue p105 human enhancer of filamentation 1 (HEF1). PTK2B is particularly enriched in pulmonary vascular endothelial cells and lung, a major organ site for tumor metastasis. Inhibitors of PTK2B may be useful in the treatment of lung cancer.
GenBank S80542
Specifications
mg/mL of enzyme in Tris/HCl pH 7.5, 150 mM NaCl, 0.1 mM EGTA, 0.03% Brij-35, 270 mM sucrose, 1 mM benzamidine, 0.2 mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution. | |
Human | |
N-terminal 6His | |
Full-length | |
Pyk2, active |
117.2 kDa | |
Recombinant | |
Recombinant enzyme expressed in Sf21 insect cells | |
≥53% |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.