Learn More
Thermo Scientific™ SOLA™ SPE Cartridges
Achieve greater reproducibility with cleaner more consistent extracts compared to conventional SPE, phospholipid removal and protein precipitation products.
Brand: Thermo Scientific™ 60409-005
2019.59 SEK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Grow your laboratory productivity. Thermo Scientific™ SOLA™ SPE Cartridges revolutionize solid phase extraction (SPE). This first fritless SPE product range enables greater sample throughput in bioanalytical laboratories where low elution volumes combined with greater reproducibility, lower failure rates and cleaner extracts are key to achieving better productivity.
More about SOLA SPE cartridges
SOLA products are available in a 2mg/1mL, 10mg/1mL and 10mg/3mL cartridge formats. They provide highly reproducible, clean and consistent sample extracts for bioanalysis by utilizing the unique award winning fritless SPE technology. The enhanced performance of SOLA cartridges gives you greater confidence in your analytical results and lowers operating costs - without compromising ease of use or requiring complex method development.
SOLA cartridges provide unparalleled performance characteristics compared to conventional SPE, phospholipid removal and protein precipitation products.
What SOLA Cartridges Mean to Your Analysis
- Prevent sample failures
- Prevent blocked wells or cartridges
- Reduced reanalysis
Specifications
SOLA WAX SPE Cartridge 30 mg/3 mL | |
Bioanalytical Applications - Strong Ion-Exchange Retention of Strong Acidic Compounds. Sorbent Charge Can be Activated or Deactivated. Complementary Reversed Phase Retention of Neutral Compounds. | |
Mixed Mode Weak Anion Exchanger, Polymeric | |
3 mL |
30 mg | |
SOLA | |
SPE Cartridge |
Product Suggestions
Customers who viewed this item also viewed.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.