missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ SR 9238
Potent and selective LXR inverse agonist
2294.76 SEK - 8855.06 SEK
Specifications
Chemical Name or Material | Ethyl 5-[[[[3′-(Methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl][(2,4,6-trimethylphenyl)sulfonyl]amino]methyl]-2-furancarboxylate |
---|---|
CAS | 1416153-62-2 |
Target | LXR-like Receptor Antagonists |
Molecular Formula | C31H33NO7S2 |
Formula Weight | 595.73 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18727408
|
Tocris Bioscience™
5854/10_TEST |
10 mg |
2294.76 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18750848
|
Tocris Bioscience™
5854/50 |
50 mg |
8855.06 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective LXR inverse agonist (IC50 values are 43 and 214 nM for LXRβ and LXRα, respectively). Selectively inhibits liver LXR over peripheral LXR. Exhibits selectivity for LXR over a panel of 20 other nuclear receptors, including FXR. Reduces hepatic steatosis in obese mice.Specifications
Ethyl 5-[[[[3′-(Methylsulfonyl)[1,1′-biphenyl]-4-yl]methyl][(2,4,6-trimethylphenyl)sulfonyl]amino]methyl]-2-furancarboxylate | |
LXR-like Receptor Antagonists | |
595.73 |
1416153-62-2 | |
C31H33NO7S2 | |
>98% |
Safety and Handling
Recommended Storage : Store at 4°C