Learn More
Swann-Morton™ Stainless Steel Surgical Scalpel Handle B3
Ergonomically designed stainless steel scalpel handle. Swann-Morton™ Stainless Steel Surgical Scalpel Handle is one of an assortment of finely crafted scalpel handles designed for precision handling.
Brand: Swann-Morton™ 0923
Description
- Ergonomic design
- Precise, finely crafted
- Suitable for a variety of procedures and blades
Specifications
Stainless Steel | |
Barron Handle | |
Blade # 6, # 9, # 10, # 10A, # 11, # 11P, # 11E, # 12, # 12D, # 13, # 14, # 15, # 15A, # 15C, # 15D, # 15T, # 16, # 40 and # SG3. |
Scalpel Handle | |
# 3 | |
Non-sterile |
Product Suggestions
Customers who viewed this item also viewed.