Learn More
VEE GEE Thermometer, Immersion with Armor
The serialized armored glass thermometer's armor protects the thermometer from rough handling and shock during use.
645.00 SEK - 825.00 SEK
Specifications
Bottle Fill | Safety Blue |
---|---|
For Use With (Application) | Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications |
Material | Glass |
Probe Type | Immersion/General Purpose (Tapered) |
Thermometer Fill | Safety Blue |
Product Code | Brand | Immersion Length | Immersion Type | Temperature Range (English) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Immersion Length | Immersion Type | Temperature Range (English) | Price | Quantity & Availability | |||
17811234
|
VEE GEE
80701-A |
30.48 cm (12 in.) | Total Immersion | -30 to +120°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17891194
|
VEE GEE
80702-A |
30.48 cm (12 in.) | Total Immersion | -4 to +230°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17821244
|
VEE GEE
80703-A |
30.48 cm (12 in.) | Total Immersion | 0 to 300°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17861244
|
VEE GEE
80706-A |
30.48 cm (12 in.) | Total Immersion | -31 to +122°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17801254
|
VEE GEE
80707-A |
30.48 cm (12 in.) | Total Immersion | -4 to +230°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17891244
|
VEE GEE
80708-A |
30.48 cm (12 in.) | Total Immersion | -4 to +302°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17851204
|
VEE GEE
80901-A |
To immersion line | Partial Immersion | -30 to +120°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17881254
|
VEE GEE
80902-A |
To immersion line | Partial Immersion | -4 to +230°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17801264
|
VEE GEE
80903-A |
To immersion line | Partial Immersion | 0 to 300°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17801214
|
VEE GEE
80905-A |
To immersion line | Partial Immersion | 20 to 500°F |
825.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17871204
|
VEE GEE
80906-A |
To immersion line | Partial Immersion | -31 to +122°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17811264
|
VEE GEE
80907-A |
To immersion line | Partial Immersion | -4 to +230°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17861194
|
VEE GEE
80908-A |
To immersion line | Partial Immersion | -4 to +302°F |
645.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
17831214
|
VEE GEE
80910-A |
To immersion line | Partial Immersion | -10 to +250°F |
825.00 SEK
Each |
Estimated Shipment: 05-02-2025
Log in to see stock. |
|||
Description
- Includes nickel-plated brass armor
- Feature Safety Blue fill against a white background for easy reading
- Accuracy is equal to one subdivision
- Immersion: Total or Partial option
Specifications
Safety Blue | |
Glass | |
Safety Blue | |
Nickel-plated brass armor |
Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications | |
Immersion/General Purpose (Tapered) | |
Serialized Glass Thermometer | |
±1 subdivision |