Learn More
VEE GEE Thermometer, Immersion with Armor
The serialized armored glass thermometer's armor protects the thermometer from rough handling and shock during use.
645.00 SEK - 825.00 SEK
Specifications
Bottle Fill Safety Blue
For Use With (Application) Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications
Material Glass
Probe Type Immersion/General Purpose (Tapered)
Thermometer Fill Safety Blue
View More Specs
Products 14
Product Code Brand Immersion Length Immersion Type Temperature Range (English) Price Quantity & Availability  
Product Code Brand Immersion Length Immersion Type Temperature Range (English) Price Quantity & Availability  
17811234
View Documents
VEE GEE
80701-A
30.48 cm (12 in.) Total Immersion -30 to +120°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17891194
View Documents
VEE GEE
80702-A
30.48 cm (12 in.) Total Immersion -4 to +230°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17821244
View Documents
VEE GEE
80703-A
30.48 cm (12 in.) Total Immersion 0 to 300°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17861244
View Documents
VEE GEE
80706-A
30.48 cm (12 in.) Total Immersion -31 to +122°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17801254
View Documents
VEE GEE
80707-A
30.48 cm (12 in.) Total Immersion -4 to +230°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17891244
View Documents
VEE GEE
80708-A
30.48 cm (12 in.) Total Immersion -4 to +302°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17851204
View Documents
VEE GEE
80901-A
To immersion line Partial Immersion -30 to +120°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17881254
View Documents
VEE GEE
80902-A
To immersion line Partial Immersion -4 to +230°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17801264
View Documents
VEE GEE
80903-A
To immersion line Partial Immersion 0 to 300°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17801214
View Documents
VEE GEE
80905-A
To immersion line Partial Immersion 20 to 500°F
825.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17871204
View Documents
VEE GEE
80906-A
To immersion line Partial Immersion -31 to +122°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17811264
View Documents
VEE GEE
80907-A
To immersion line Partial Immersion -4 to +230°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17861194
View Documents
VEE GEE
80908-A
To immersion line Partial Immersion -4 to +302°F
645.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
17831214
View Documents
VEE GEE
80910-A
To immersion line Partial Immersion -10 to +250°F
825.00 SEK
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
Description
Description
  • Includes nickel-plated brass armor
  • Feature Safety Blue fill against a white background for easy reading
  • Accuracy is equal to one subdivision
  • Immersion: Total or Partial option
Specifications
Specifications
Safety Blue
Glass
Safety Blue
Nickel-plated brass armor
Laboratories, universities, food/beverage, environmental, wastewater and many other general-purpose applications
Immersion/General Purpose (Tapered)
Serialized Glass Thermometer
±1 subdivision