Learn More
Tirbanibulin Mesylate, MedChemExpress
MedChemExpress Tirbanibulin Mesylate (KX2-391 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.
Brand: Medchem Express HY-10340B
Description
Tirbanibulin Mesylate (KX2-391 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.Specifications
Tirbanibulin Mesylate | |
Off-White | |
C27H33N3O6S | |
KX2-391 MesylateKX01 Mesylate | |
O=C(NCC1=CC=CC=C1)CC2=NC=C(C3=CC=C(OCCN4CCOCC4)C=C3)C=C2.CS(=O)(O)=O | |
527.63 | |
Research | |
Cancer-Kinase/protease |
1080645-95-9 | |
Research | |
10 mg | |
DMSO : 100 mg/mL (189.53 mM; Need ultrasonic) | |
527.63 | |
99.97% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only