Learn More
Tirbanibulin Mesylate, MedChemExpress
MedChemExpress Tirbanibulin Mesylate (KX2-391 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.
1042.56 SEK - 8527.21 SEK
Specifications
Chemical Name or Material | Tirbanibulin Mesylate |
---|---|
CAS | 1080645-95-9 |
Color | Off-White |
Purity Grade Notes | Research |
Molecular Formula | C27H33N3O6S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16494876
![]() |
Medchem Express
HY-10340B |
100 mg |
8527.21 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16456567
![]() |
Medchem Express
HY-10340B |
5 mg |
1042.56 SEK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16430217
![]() |
Medchem Express
HY-10340B |
50 mg |
4979.39 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16403848
![]() |
Medchem Express
HY-10340B |
10 mg |
1291.53 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tirbanibulin Mesylate (KX2-391 Mesylate) is an inhibitor of Src that targets the peptide substrate site of Src, with GI50 of 9-60 nM in cancer cell lines.Specifications
Tirbanibulin Mesylate | |
Off-White | |
C27H33N3O6S | |
DMSO : 100 mg/mL (189.53 mM; Need ultrasonic) | |
527.63 | |
99.97% | |
Solid |
1080645-95-9 | |
Research | |
KX2-391 MesylateKX01 Mesylate | |
O=C(NCC1=CC=CC=C1)CC2=NC=C(C3=CC=C(OCCN4CCOCC4)C=C3)C=C2.CS(=O)(O)=O | |
527.63 | |
Research | |
Cancer-Kinase/protease |
Safety and Handling
missing translation for 'shelfLife' : -20°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
Recommended Storage : -20°C, protect from light, stored under nitrogen∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light, stored under nitrogen)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.